missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 1/4 in. Ceramic Spheres

Use to break hard/brittle samples. Specialty applications for the 1/4 in. Ceramic Spheres include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.

$240.47 - $2698.50

Specifications

Bead Type Zirconium oxide coated ceramic
Content And Storage Room Temperature
Sample Type Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores
For Use With (Application) Break hard/brittle samples
Product Type Bulk Matrix
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP116540034
View Documents
MP Biomedicals™
116540034
50 Pk.
Pack of 50 Each for $240.47
Only null left
 
116540422
View Documents
MP Biomedicals™
116540422
1000 Pk.
Each for $2,698.50
Only null left
 
116540424
View Documents
MP Biomedicals™
116540424
100 Pk.
Each for $311.17
Only null left
 
116540412
View Documents
MP Biomedicals™
116540412
500 Pk.
Each for $1,440.31
Only null left
 
Description

Description

  • Zirconium Oxide-coated ceramic grinding spheres
  • Chemically inert
  • Component of Lysing Matrix A and M tubes
Specifications

Specifications

Zirconium oxide coated ceramic
Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores
Bulk Matrix
1/4 in. Zirconium Oxide Banded Satellite for Grinding
Room Temperature
Break hard/brittle samples
Unlimited
Videos
SDS
Documents

Documents

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.