missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 1/4 in. Ceramic Spheres

Catalog No. 116540422
Change view
Click to view available options
Quantity:
100 Pk.
1000 Pk.
50 Pk.
500 Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
116540422 1000 Pk.
MP116540034 50 Pk.
116540424 100 Pk.
116540412 500 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 116540422 Supplier MP Biomedicals™ Supplier No. 116540422
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use to break hard/brittle samples. Specialty applications for the 1/4 in. Ceramic Spheres include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.

  • Zirconium Oxide-coated ceramic grinding spheres
  • Chemically inert
  • Component of Lysing Matrix A and M tubes

Specifications

Bead Type Zirconium oxide coated ceramic
Content And Storage Room Temperature
Sample Type Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores
For Use With (Application) Break hard/brittle samples
Product Type Bulk Matrix
Quantity 1000 Pk.
Sample Volume Unlimited
Synonym 1/4 in. Zirconium Oxide Banded Satellite for Grinding

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.