Learn More
SOMNI Scientific™ MouseTaxic™ Stereotaxic Nose Cone For Active Waste Gas Scavenging
Catalog No. 04777143
Change view
Click to view available options
Includes:
Supply tubing,connectors
Supply tubing,connectors,head clamp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
04777143 Supply tubing,connectors,head clamp
04777141 Supply tubing,connectors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04777143 Supplier SOMNI Scientific™ Supplier No. NC19103
Designed to be used with stereotaxic devices to safely deliver and actively scavenge inhalant anesthesia
  • Mouse size
  • Stereotaxic nosecone
  • Use with attenuated active scavenging
  • Supply tubing and connectors included
  • Contact SOMNI Scientific to check compatibility
  • Works with SOMNI EPS-3 or attenuated in-house vacuum system
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
For Use With (Equipment) AMD-17106, AMD-17004, TT-17000, TT-17001, TT-17002, WG-15001, WG-15101
Material 3D Printed
Length (Metric) 8.3 cm
Width (Metric) 5.7 cm
Includes Supply tubing,connectors,head clamp
For Veterinary Use Only