missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse Th17 Cytokine Staining Panel

Catalog No. 501125225
Change view
Click to view available options
Detects:
IL-17A, IL-17F, IL-21, IL-22
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detects
501125225 IL-17A, IL-17F, IL-21, IL-22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125225 Supplier Invitrogen™ Supplier No. 88841144

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Anti-Mouse IL-17A (eBio17B7) FITC : 25 ug ([optimal] = 0.25 μg)*
  • Anti-Mouse IL-17F (eBio18F10) PE : 25 ug ([optimal] = 0.5 μg)*
  • Anti-Mouse IL-22 (1H8PWSR) PerCP-eFluor 710 : 25 ug ([optimal] = 0.125 μg)*
  • Anti-Mouse IL-21 (FFA21) APC : 25 ug ([optimal] = 0.125 μg)*
  • Anti-Mouse CD4 (RM4-5) eFluor 450 : 25 ug ([optimal] = 0.125 μg)*
  • IC Fixation Buffer
  • Permeabilization Buffer (10X)
  • *The optimal concentration is based on 1 million cells in a 100 μL total staining volume.

Includes all reagents needed for simultaneous flow cytometric detection of all the major cytokines produced by the Th17 lineage (IL-17A, IL-17F, IL-21 and IL-22).

Also included: An anti-CD4 antibody that can be used after fixation and permeabilization, as well as IC Fixation and Permeabilization Buffers.

Specifications

Detects IL-17A, IL-17F, IL-21, IL-22
For Use With (Application) Intracellular Staining Followed by Flow Cytometric Analysis
Storage Buffer Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Content And Storage 2°C to 8°C. Do not freeze. Light sensitive material.

For Research Use Only