Recombinant Protein;100UG;Mouse Neuropilin-1, His Tag (NR1-M52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Pro 856 (Accession # P97333-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 95.5 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human VEGF165, Tag Free (MALS verified) (Cat. No. VE5-H4210) at 2 g/mL (100 L/well) can bind Mouse Neuropilin-1, His Tag (Cat. No. NR1-M52H3) with a linear range of 2-10 ng/mL (QC tested).