Mouse Jagged 1 (JAG1) ELISA Kit for detecting JAG1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QXX0. Gene ID: 16449. Target Synonyms: CD339; AGS; AHD; AWS; HJ1; JAGL1; Alagille Syndrome; Jagged 1 Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.