missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-4 Recombinant Protein

Catalog No. ENRMIL4I
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENRMIL4I 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENRMIL4I Supplier Invitrogen™ Supplier No. RMIL4I

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant Protein

Mature mouse IL-4 was expressed in Pichia pastoris, X-33. GenBank Accession # P07750, amino acids 21 - 140. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: HIHGCDKNHL. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity of recombinant mouse IL-4 was determined by measuring the proliferation of mouse HT-2 cells. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 0.5 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.

Specifications

Accession Number P07750
For Use With (Application) Bioactivity
Formulation PBS with no preservative
Gene ID (Entrez) 16189
Molecular Weight (g/mol) 13.6 kDa
Name Mouse IL-4
Purification Method Purified
Quantity 20 μg
Storage Requirements 4°C
Regulatory Status RUO
Gene Alias B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra
Common Name IL-4
Gene Symbol Il4
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Mouse IL-4, amino acids 21-140 (Accession # P07750)
Activity ED50 = 3.3-4.9 ng/mL: Determined by the proliferation of mouse HT-2 cells.
Expression System P. pastoris
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less