missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse Hematopoietic Lineage Biotin Panel

Catalog No. 501125212
Change view
Click to view available options
Quantity:
1000 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125212 1000 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Azide
Catalog No. 501125212 Supplier Invitrogen™ Supplier No. 88777475

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Azide

Includes

  • Biotin anti-mouse CD3 (145-2C11): 2ml
  • Biotin anti-mouse CD45R/B220 (RA3-6B2): 2ml
  • Biotin anti-mouse CD11b (M1/70): 2ml
  • Biotin anti-mouse Erythroid marker (TER-119): 2ml
  • Biotin anti-mouse Ly-6G (RB6-8C5): 2ml

Contains 5 biotinylated antibodies that can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on their cell surface antigen expression.

The eBioscience Mouse Hematopoietic Lineage Flow Panel contains 5 biotinylated antibodies that can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on their cell surface antigen expression.

Reactivity/Species
Mouse

Reported Application
Flow Cytometric Analysis

Specifications

Content And Storage 2° to 8°C
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
No. of Tests 1000 tests
Quantity 1000 tests
For Use With (Application) Flow Cytometry
Product Line eBioscience
Product Type Biotin Panel

For Research Use Only. Not for use in diagnostic procedures.