missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse EGF Native Protein

Numéro de catalogue. 53003018
Encompass
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
53003018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 53003018 Fournisseur Gibco™ Code fournisseur 53003018
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Native Protein

Reconstitution: Reconstituted EGF should be aliquoted and stored in polypropylene vials and is stable at -5°C to -20°C for up to two months. Avoid repeated freezing and thawing. Stability of dilute solutions of EGF may be prolonged by addition of bovine serum albumin to a final concentration of 1.0%. This natural preparation of murine epidermal growth factor (EGF) was isolated from male mouse submaxillary glands. It is not sterile as provided.

EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.

Spécifications

Numéro d'enregistrement P01132
À utiliser avec (application) Bioactivity
Formule Protein with no preservative
ID du gène (Entrez) 13645
Poids moléculaire (g/mol) 6 kDa
Nom Mouse EGF
Méthode de purification Purified
Quantité 100 μg
Conditions de stockage -20°C
Statut réglementaire RUO
Alias du gène AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone
Nom usuel EGF
Symbole de gène EGF
Type de produit Protein
Conjugué Unconjugated
Espèces Mouse
Contenu et stockage -20°C
Shipping condition (Condition d'expédition) Approved for shipment at Room Temperature or on Wet or Dry Ice
Famille de protéines Growth Factors & Receptors
Forme de protéine Native, Ligand
Forme Lyophilized
Sous-type de protéine EGF (Epidermal Growth Factor)
Catégorie de recherche Neurobiology, Stem Cell Research, Differentiation, Oncology
Grade de qualité ou de pureté >95% by SDS-PAGE
Afficher plus de résultats Afficher moins de résultats