Learn More
Invitrogen™ Mouse anti-Bovine Ig Mix Secondary Antibody
Mouse Monoclonal Secondary Antibody
Supplier:  Invitrogen™ SA110639
 View more versions of this product
Catalog No. 01-671-606
Only null left
Explore available promotions
Description
Description
SA1-10639 detects bovine Immunoglobulin. SA1-10639 has been successfully used in ELISA applications. The SA1-10639 immunogen is purified bovine immunoglobulin.
Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Bovine Ig Mix
Monoclonal
Unconjugated
Mouse
Protein A
RUO
Bovine
Secondary Antibody
IgG1
ELISA
BIG10-123.1
PBS with no preservative; pH 7.4
Purified bovine immunoglobulin.
500 μg
Secondary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions