missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Bovine Ig Mix Secondary Antibody

Catalog No. p-7244519
Change view
Click to view available options
Quantity:
100 μg
500 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01671606 500 μg
PISA510360 1 mg
PISA510361 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01671606 Supplier Invitrogen™ Supplier No. SA110639
Only null left

Mouse Monoclonal Secondary Antibody

SA1-10639 detects bovine Immunoglobulin. SA1-10639 has been successfully used in ELISA applications. The SA1-10639 immunogen is purified bovine immunoglobulin.

Anti-Bovine secondary antibodies are affinity-purified antibodies with well-characterized specificity for bovine (cow, ox) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Bovine Ig Mix
Applications ELISA
Classification Monoclonal
Clone BIG10-123.1
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Host Species Mouse
Immunogen Purified bovine immunoglobulin.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less