ACROBiosystems Inc Anti-Adalimumab Antibody (AY23b) (recommended for PK/PD)
Catalog No. 502719939
Adalimumab is a human monoclonal antibody against TNF-alpha. It is produced by recombinant DNA technology using a mammalian cell expression system. It consists of 1330 amino acids and has a molecular weight of approximately 148 kilodaltons.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Adalimumab is a human monoclonal antibody against TNF-alpha. It is produced by recombinant DNA technology using a mammalian cell expression system. It consists of 1330 amino acids and has a molecular weight of approximately 148 kilodaltons.