missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Mounted Human Skeleton, Half Size

Catalog No. S15013
Change view
Click to view available options
Skeleton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S15013 Skeleton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15013 Supplier Eisco™ Supplier No. AMCHA105

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Half Size Mounted Human Skeleton exhibits realistic details making it ideal for educational study, as well as for patient demonstrations.

  • Quality hardware allows for easy movement of major joints
  • Mandible is articulated and removable for closer study of jaw, teeth, and internal structures
  • Arms and legs can be removed for storage and study
  • Features realistic details, textures, and bony landmarks
  • Half sized human skeleton stands 34 in. tall
  • Mounted on base with metal rod and sturdy stand
  • Made of durable and sturdy plastisol polymer
  • Skeleton is washable and easy to clean to hold up to years of handling
  • Includes key card for identifying and studying the various bones that make up a full skeleton

Specifications

Color White
For Use With (Application) Educational Study, Patient Demonstrations
Height (English) 34 in.
Height (Metric) 86 cm
Includes English Key Card
Material Plastisol Polymer
Mounting Stand with Rod
Product Type Model, Skeleton