missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Mortar and Pestle Set

Features a flat bottom, unglazed grinding surfaces, and spout for easy pouring. Eisco™ Mortar and Pestle Set is made from lab quality porcelain and designed for efficient and uniform grinding of various materials.

Supplier:  Eisco™ CH0540F

 Afficher plus de versions de ce produit

Numéro de catalogue. S39828


Il en reste null

Description

Description

  • Made from lab quality porcelain
  • Outside of mortar and handle of pestle is glazed
  • Inner bowl and pestle end are not glazed
  • Spout for easing pouring
  • Flat bottom for stability
Spécifications

Identifiants chimiques

Mortar and Pestle
Porcelain

Spécifications

1500 mL
Autoclavable
8.25 in.
Mortar and Pestle
Grinding
12 cm
19.7 cm
Thick Wall
Glazed Outer
50 fl oz
210 mm
Porcelain
4.75 in.
Mortar, Pestle
7.75 in.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions