missing translation for 'onlineSavingsMsg'
Learn More

Mordant Orange 1 98.0+%, TCI America™

Catalog No. A057925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A057925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A057925G Supplier TCI America Supplier No. A057925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2243-76-7
Molecular Formula C13H9N3O5
Molecular Weight (g/mol) 287.23
MDL Number MFCD00007313
InChI Key RTQYUQUDISIJCV-UHFFFAOYSA-N
Synonym Alizarin Yellow R, 5-(4-Nitrophenylazo)salicylic Acid
PubChem CID 6364549
IUPAC Name 3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylic acid
SMILES OC(=O)C1=CC(C=CC1=O)=NNC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Quantity 25 g
Formula Weight 309.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mordant Orange 1
EINECSNumber (5)-2054
RTECSNumber VO5310000
TSCA Yes