missing translation for 'onlineSavingsMsg'
Learn More

MOPSO, 0.2M buffer soln., pH 7.0, Thermo Scientific™

Catalog No. AAJ61469AK
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61469AK 250 mL
AAJ61469AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61469AK Supplier Thermo Scientific Chemicals Supplier No. J61469AK
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 79803-73-9
Molecular Formula C7H14NNaO5S
Molecular Weight (g/mol) 247.241
MDL Number MFCD00070007
InChI Key WSFQLUVWDKCYSW-UHFFFAOYSA-M
PubChem CID 23675131
IUPAC Name sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+]

Specifications

Concentration 0.2 M
pH 7.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material MOPSO buffer soln.
Hazard Category H335
TSCA No
Recommended Storage Ambient temperatures