missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MOPS, Sodium, ULTROL™ Grade, Calbiochem™,
Specifications
Specifications
| Quantity | 1 kg |
| Assay Percent Range | ≥98.0% |
| Solubility Information | Soluble in cold water |
| Formula Weight | 231.2 |
| Grade | by titration (dry basis) |
| Chemical Name or Material | zwitterionic buffer |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F). |