missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MOPS, Free Acid, ULTROL™ Grade, Calbiochem™,
A zwitterionic buffer useful in the pH range of 6.5 to 7.9
$174.00 - $825.00
Chemical Identifiers
| CAS | 1132-61-2 |
|---|---|
| Molecular Formula | C7H15NO4S |
| Molecular Weight (g/mol) | 209.26 |
| MDL Number | MFCD00006183 |
| InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
| Synonym | 3-(N-Morpholino)propanesulfonic Acid |
| PubChem CID | 70807 |
| ChEBI | CHEBI:44115 |
| IUPAC Name | 4-(3-sulfonatopropyl)morpholin-4-ium |
| SMILES | [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
475898100GM
|
MilliporeSigma™
475898100GM |
100 g |
Each for $174.00
|
|
|||||
|
475898500GM
|
MilliporeSigma™
475898500GM |
500 g |
Each for $584.00
|
|
|||||
|
4758981KG
|
MilliporeSigma™
4758981KG |
1 kg |
Each for $825.00
|
|
|||||
Chemical Identifiers
| 1132-61-2 | |
| 209.26 | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| 70807 | |
| 4-(3-sulfonatopropyl)morpholin-4-ium |
| C7H15NO4S | |
| MFCD00006183 | |
| 3-(N-Morpholino)propanesulfonic Acid | |
| CHEBI:44115 | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
Specifications
| 1132-61-2 | |
| ≥99.0% | |
| MFCD00006183 | |
| Soluble in water | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 | |
| 209.26 | |
| CHEBI:44115 | |
| by titration |
| 100 g | |
| C7H15NO4S | |
| 3-(N-Morpholino)propanesulfonic Acid | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| 4-(3-sulfonatopropyl)morpholin-4-ium | |
| 70807 | |
| 209.3 | |
| zwitterionic buffer |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze).