missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MOPS, Free Acid, ULTROL™ Grade, Calbiochem™,

A zwitterionic buffer useful in the pH range of 6.5 to 7.9

Supplier:  MilliporeSigma™ 475898500GM

 View more versions of this product

Catalog No. 475898500GM


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

zwitterionic buffer
500 g
≥99.0%
MFCD00006183
Soluble in water
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
ULTROL
1132-61-2
by titration
C7H15NO4S
3-(N-Morpholino)propanesulfonic Acid
DVLFYONBTKHTER-UHFFFAOYSA-N
4-(3-sulfonatopropyl)morpholin-4-ium
70807
209.3

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
4-(3-sulfonatopropyl)morpholin-4-ium
C7H15NO4S
MFCD00006183
3-(N-Morpholino)propanesulfonic Acid
CHEBI:44115
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : 15°C to 30°C (Do not freeze).

SDS
Documents

Documents

Promotions

Promotions