missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals Inc MOPS, free acid, 100.2%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219483725

 View more versions of this product

Catalog No. ICN19483725



Description

Description

Specifications

Specifications

MOPS
25 g
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
4-(3-sulfonatopropyl)morpholin-4-ium
70807
209.3
1132-61-2
C7H15NO4S
3-[N-Morpholino]propanesulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
1.002

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
C7H15NO4S
MFCD00006183
3-[N-Morpholino]propanesulfonic acid
CHEBI:44115
SDS
Documents

Documents

Promotions

Promotions