missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, free acid, 100.2%, MP Biomedicals™
$83.65 - $83.65
Chemical Identifiers
| CAS | 1132-61-2 |
|---|---|
| Molecular Formula | C7H15NO4S |
| Molecular Weight (g/mol) | 209.26 |
| MDL Number | MFCD00006183 |
| InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
| Synonym | 3-[N-Morpholino]propanesulfonic acid |
| PubChem CID | 70807 |
| ChEBI | CHEBI:44115 |
| SMILES | [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19483725
|
MP Biomedicals Inc
0219483725 |
25 g |
Each for $83.65
|
|
|||||
Chemical Identifiers
| 1132-61-2 | |
| 209.26 | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| 70807 | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
| C7H15NO4S | |
| MFCD00006183 | |
| 3-[N-Morpholino]propanesulfonic acid | |
| CHEBI:44115 |
Specifications
| 1132-61-2 | |
| C7H15NO4S | |
| 3-[N-Morpholino]propanesulfonic acid | |
| [O-]S(=O)(=O)CCC[NH+]1CCOCC1 | |
| 209.26 | |
| CHEBI:44115 | |
| 1.002 |
| 25 g | |
| MFCD00006183 | |
| DVLFYONBTKHTER-UHFFFAOYSA-N | |
| 4-(3-sulfonatopropyl)morpholin-4-ium | |
| 70807 | |
| 209.3 | |
| MOPS |