missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 99%

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

$73.99 - $623.26

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 4-Morpholinepropanesulfonic acid; 3-(4-Morpholinyl)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1291414
View Documents
Thermo Scientific Chemicals
A1291414
25 g
Each for $73.99
Only null left
 
AAA1291422
View Documents
Thermo Scientific Chemicals
A1291422
100 g
Each for $162.78
Only null left
 
AAA1291436
View Documents
Thermo Scientific Chemicals
A1291436
500 g
Each for $623.26
Only null left
 
Description

Description

MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

Solubility
Soluble in water.    

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
C7H15NO4S
MFCD00006183
4-Morpholinepropanesulfonic acid; 3-(4-Morpholinyl)propanesulfonic acid
CHEBI:44115

Specifications

277°C to 280°C (decomposition)
25 g
99%
MFCD00006183
14,6265
Soluble in water.
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
99%
1132-61-2
110°C (230°F)
C7H15NO4S
1106776
4-Morpholinepropanesulfonic acid; 3-(4-Morpholinyl)propanesulfonic acid
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
209.27
MOPS
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-478-5

RTECSNumber : QE9104530

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only