missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 99%

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

Supplier:  Thermo Scientific Chemicals A1291414

 View more versions of this product

Catalog No. AAA1291414


Only null left

Description

Description

MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

Solubility
Soluble in water.    

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

MOPS
1132-61-2
110°C (230°F)
C7H15NO4S
1106776
4-Morpholinepropanesulfonic acid; 3-(4-Morpholinyl)propanesulfonic acid
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
209.27
277°C to 280°C (decomposition)
25 g
99%
MFCD00006183
14,6265
Soluble in water.
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
99%

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
C7H15NO4S
MFCD00006183
4-Morpholinepropanesulfonic acid; 3-(4-Morpholinyl)propanesulfonic acid
CHEBI:44115
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-478-5

RTECSNumber : QE9104530

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only