missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 99%, for biochemistry

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

$88.16 - $750.20

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-Morpholino)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC172630250
SDS View Documents
Thermo Scientific Chemicals
172630250
25 g Plastic bottle
Each for $88.16
Only null left
 
AC172631000
SDS View Documents
Thermo Scientific Chemicals
172631000
100 g Plastic bottle
Each for $203.10
Only null left
 
AC172635000
SDS View Documents
Thermo Scientific Chemicals
172635000
500 g Plastic bottle
Each for $750.20
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
C7H15NO4S
MFCD00006183
3-(N-Morpholino)propanesulfonic acid
CHEBI:44115

Specifications

(1 M aq. soln.) (1 cm cell) at 0nm
1132-61-2
100.0
>277°C
Crystalline Powder
99%
C7H15NO4S
Plastic bottle
3-(N-Morpholino)propanesulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
≥98.5%
MOPS, 99%
(1M aq. soln.) (1 cm cell)
98.5
White
3.0 to 4.5 (1% aq. soln. at 25°C)
25 g
Authentic
MFCD00006183
15, 6352
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
209.26
Biochemical
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Emergency Overview
Causes eye, skin, and respiratory tract irritation.
NFPA
Health: 2
Flammability:

Warning

EINECSNumber : 214-478-5

RTECSNumber : QE9104530

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only