missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 99%, for biochemistry

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

Supplier:  Thermo Scientific Chemicals 172635000

 View more versions of this product

Catalog No. AC172635000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Specifications

0.03 max. at 280nm
99%
277.0°C to 282.0°C
98.5
White
3.0 to 4.5 (1% aq. soln. at 25°C)
500 g
99%
C7H15NO4S
Plastic bottle
3-(N-Morpholino)propanesulfonic acid
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
209.26
Biochemical
MOPS
(1M aq. soln.) (1 cm cell)
1132-61-2
100.0
>277°C
Crystalline Powder
116°C
Authentic
MFCD00006183
15, 6352
Solubility in water: 1000g/L (20°C). Other solubilities: <10g/L ethanol (20°C)
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
99%

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
C7H15NO4S
MFCD00006183
3-(N-Morpholino)propanesulfonic acid
CHEBI:44115
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 214-478-5

RTECSNumber : QE9104530

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only