missing translation for 'onlineSavingsMsg'
Learn More

MOPS, ≥98.5% (Dry Basis), Ultrapure

Catalog No. AAJ19256A1
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ19256A1 1 kg Plastic bottle
AAJ1925622 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ19256A1 Supplier Thermo Fisher Scientific Supplier No. J19256A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-[N-Morpholino]propane sulfonic Acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES C1COCCN1CCCS(=O)(=O)O

Specifications

Melting Point 277°C to 280°C (decomposition)
Physical Form Crystalline Powder
Quantity 1 kg
Flash Point 110°C
Linear Formula HO3S(CH2)3(C4H8NO)
Packaging Plastic bottle
Beilstein 1106776
Formula Weight 209.26
Percent Purity ≥98.5%
Assay Dry basis
Chemical Name or Material MOPS
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-478-5
RTECSNumber QE9104530
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only