missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 101%, MP Biomedicals™

Catalog No. ICN19067083
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19067083 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19067083 Supplier MP Biomedicals Inc Supplier No. 0219067083
Only null left

Chemical Identifiers

CAS 71119-22-7
Molecular Formula C7H14NNaO4S
Molecular Weight (g/mol) 231.242
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym 3-[N-Morpholino]propanesulfonic acid
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 1.01
Formula Weight 231.2
Chemical Name or Material MOPS