missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 0.2M buffer soln., pH 7.4, low endotoxin, Thermo Scientific™

Catalog No. AAJ67497AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67497AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67497AE Supplier Thermo Scientific Chemicals Supplier No. J67497AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Color Colorless
Concentration 0.2 M
pH 7.4
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material MOPS, low endotoxin
Hazard Category H335
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold