missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monosodium 5-Sulfoisophthalate 98.0+%, TCI America™
Supplier: TCI America S014825G
Specifications
| Monosodium 5-Sulfoisophthalate | |
| White | |
| MFCD00007495 | |
| 5-sulfafe-3-phthalicacid | |
| C1=C(C=C(C=C1C(=O)O)S(=O)(=O)O)C(=O)[O-].[Na+] | |
| 268.171 | |
| 268.17 | |
| Crystalline Powder |
| 6362-79-4 | |
| C8H5NaO7S | |
| 25 g | |
| YXTFRJVQOWZDPP-UHFFFAOYSA-M | |
| sodium;3-carboxy-5-sulfobenzoate | |
| 57488606 | |
| ≥98.0% (T) |
Chemical Identifiers
| 6362-79-4 | |
| 268.171 | |
| YXTFRJVQOWZDPP-UHFFFAOYSA-M | |
| 57488606 | |
| C1=C(C=C(C=C1C(=O)O)S(=O)(=O)O)C(=O)[O-].[Na+] |
| C8H5NaO7S | |
| MFCD00007495 | |
| 5-sulfafe-3-phthalicacid | |
| sodium;3-carboxy-5-sulfobenzoate |
Safety and Handling
EINECSNumber : (3)-3168
RTECSNumber : DB6030000
TSCA : Yes