accessibility menu, dialog, popup

UserName

CAS RN 6362-79-4

IUPAC Name: sodium 3,5-dicarboxybenzene-1-sulfonate
Synonyms: sodium 3,5-dicarboxybenzenesulfonate
Molecular Weight (g/mol): 268.17
Molecular Formula: C8H5NaO7S
InChi Key: YXTFRJVQOWZDPP-UHFFFAOYSA-M
SMILES: [Na+].OC(=O)C1=CC(=CC(=C1)C(O)=O)S([O-])(=O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)