missing translation for 'onlineSavingsMsg'
Learn More

Monosodium 5-Sulfoisophthalate 98.0+%, TCI America™

Catalog No. p-7135834
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S014825G 25 g
S0148100G 100 g
S0148500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S014825G Supplier TCI America Supplier No. S014825G
Only null left

Chemical Identifiers

CAS 6362-79-4
Molecular Formula C8H5NaO7S
Molecular Weight (g/mol) 268.171
MDL Number MFCD00007495
InChI Key YXTFRJVQOWZDPP-UHFFFAOYSA-M
Synonym 5-sulfafe-3-phthalicacid
PubChem CID 57488606
IUPAC Name sodium;3-carboxy-5-sulfobenzoate
SMILES C1=C(C=C(C=C1C(=O)O)S(=O)(=O)O)C(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 268.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monosodium 5-Sulfoisophthalate
EINECSNumber (3)-3168
RTECSNumber DB6030000
TSCA Yes