missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Adipate 97.0+%, TCI America™

Catalog No. A173825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A173825G 25 g
A1738100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A173825G Supplier TCI America Supplier No. A173825G

Chemical Identifiers

CAS 627-91-8
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004418
InChI Key UOBSVARXACCLLH-UHFFFAOYSA-N
Synonym monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester
PubChem CID 12328
ChEBI CHEBI:70855
IUPAC Name 6-methoxy-6-oxohexanoic acid
SMILES COC(=O)CCCCC(O)=O

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 162°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Monomethyl Adipate
EINECSNumber (2)-0861
TSCA Yes