missing translation for 'onlineSavingsMsg'
Learn More

Monolaurin 98.0+%, TCI America™

Catalog No. G00815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G00815G 5 g
G008125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G00815G Supplier TCI America Supplier No. G00815G

Chemical Identifiers

CAS 142-18-7
Molecular Formula C15H30O4
Molecular Weight (g/mol) 274.401
MDL Number MFCD00037815
InChI Key ARIWANIATODDMH-UHFFFAOYSA-N
Synonym monolaurin, 1-monolaurin, lauricidin, glyceryl laurate, glyceryl monolaurate, 1-glyceryl laurate, glycerol 1-laurate, 1-monolauroyl-rac-glycerol, 1-monododecanoylglycerol, glycerol monolaurate
PubChem CID 14871
ChEBI CHEBI:75539
IUPAC Name 2,3-dihydroxypropyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(CO)O

Specifications

Melting Point 63°C
Color White
Boiling Point 186°C
Quantity 5 g
Formula Weight 274.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Monolaurin
EINECSNumber (2)-0669
RTECSNumber OF1600000
TSCA Yes