missing translation for 'onlineSavingsMsg'
Learn More

Monoethanolamine Hydrochloride, Approx. 98%, Spectrum™ Chemical

Catalog No. 18604677
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604677 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604677 Supplier Spectrum Chemical Mfg Cor Supplier No. M145625GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1456, 2002-24-6, C2H7NO·HCl

Spectrum™ Chemical Monoethanolamine Hydrochloride , also known as ethanolamine or 2-aminoethanol, is an organic chemical compound used as feedstock in the production of emulsifiers, detergents, and pharmaceuticals. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 2002-24-6
Molecular Formula C2H8ClNO
Molecular Weight (g/mol) 97.54
InChI Key PMUNIMVZCACZBB-UHFFFAOYSA-N
IUPAC Name hydrogen 2-aminoethan-1-ol chloride
SMILES [H+].[Cl-].NCCO

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity ∼98%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Monoethanolamine Hydrochloride