missing translation for 'onlineSavingsMsg'
Learn More

mono-Methyl isophthalate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 398230050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC398230050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1877-71-0
180.16
WMZNGTSLFSJHMZ-UHFFFAOYSA-N
601880
COC(=O)C1=CC=CC(=C1)C(=O)O
C9H8O4
MFCD00029972
3-methoxycarbonyl benzoic acid, mono-methyl isophthalate, methyl hydrogen isophthalate, isophthalic acid monomethyl ester, monomethyl isophthalate, isophthalic acid methyl ester, isophthalic acid, methyl ester, isophthalic acid, monomethyl ester, methyl m-phthalate, methyl 3-carboxybenzoate
3-methoxycarbonylbenzoic acid

Specifications

mono-Methyl isophthalate
96.0
Authentic
Glass bottle
(COOH)C6H4COOCH3
MFCD00029972
WMZNGTSLFSJHMZ-UHFFFAOYSA-N
3-methoxycarbonylbenzoic acid
601880
97%
1877-71-0
100.0
96% min. (GC)
C9H8O4
5g
3-methoxycarbonyl benzoic acid, mono-methyl isophthalate, methyl hydrogen isophthalate, isophthalic acid monomethyl ester, monomethyl isophthalate, isophthalic acid methyl ester, isophthalic acid, methyl ester, isophthalic acid, monomethyl ester, methyl m-phthalate, methyl 3-carboxybenzoate
COC(=O)C1=CC=CC(=C1)C(=O)O
180.16
180.16
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents