missing translation for 'onlineSavingsMsg'
Learn More

Mometasone Furoate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015659
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015659 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015659 Supplier Merck Emd Millipore Supplier No. PHR1400500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mometasone Furoate is a medication with anti-inflammatory and antipruritic properties, commonly used to treat mixed infections of the skin. It is also used as a corticosteroid for the treatment of asthma and its mode of action involves reducing the inflammation of the airways and preventing the loss of lung functions.

Specifications

CAS 83919-23-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C27H30 Cl2O6
MDL Number MFCD00866003
Synonym Mometasone furoate; (11 beta,16 alpha)-9,21-Dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione
Type Certified Reference Material
Quantity 500 mg
Formula Weight 521.43
Grade Certified Reference Material