missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum foil, 0.5mm (0.02in) thick, 99.95% (metals basis)

Catalog No. AA10045HX
Change view
Click to view available options
Quantity:
100 x 100 mm
150 x 300 mm
300 x 600 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10045HX 300 x 600 mm
AA10045GH 100 x 100 mm
AA10045HW 150 x 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10045HX Supplier Thermo Scientific Chemicals Supplier No. 010045HX

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7439-98-7 | Mo | 95.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 300 x 600 mm
Weight ≈51g/100x100mm
Form Foil
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Molybdenum foil, 0.5mm (0.02 in.) thick
EINECSNumber 231-107-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only