missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Mo), LabChem™
Supplier: LabChem LC174001
Specifications
| Molybdenum AA Standard | |
| Poly Bottle | |
| 99.6,0.4 | |
| 1g/mL | |
| 1000 ppm ±10 ppm (1mL = 1mg Mo) | |
| Colorless | |
| 500 mL | |
| Certified |
| 1000ppm (1mL = 1mg Mo) | |
| 12054-85-2 , 7732-18-5 | |
| Passes Test | |
| Liquid | |
| (NH4)6Mo7O24·4H2O | |
| Soluble in water | |
| 1235.86 |
Chemical Identifiers
| 12054-85-2 |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature