missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Mo), LabChem™

Catalog No. p-5388670
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC174007 125 mL Poly Bottle
LC174001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC174007 Supplier LabChem Supplier No. LC174007
Only null left

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5

Specifications

Packaging Poly Bottle
CAS Max % 99.6, 0.4
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg Mo)
Linear Formula (NH4)6Mo7O24·4H2O
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 1235.86
Grade Certified
Chemical Name or Material Molybdenum AA Standard, 1000ppm (1mL = 1mg Mo)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature