missing translation for 'onlineSavingsMsg'
Learn More

Molybdenite, naturally occurring mineral, grains, approximately 0.06-0.19in

Used for grain mounts, XRD and microprobe standards, as well as identification of unknown minerals. | CAS: 1309-56-4 | MoS2 | 160.07 g/mol

$49.51 - $49.51

Chemical Identifiers

CAS 1309-56-4
Molecular Formula MoS2
Molecular Weight (g/mol) 160.07
MDL Number MFCD00003470
InChI Key CWQXQMHSOZUFJS-UHFFFAOYSA-N
Synonym molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
PubChem CID 14823
ChEBI CHEBI:30704
IUPAC Name bis(sulfanylidene)molybdenum
SMILES S=[Mo]=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4264203
View Documents
Thermo Scientific Chemicals
4264203
1 g N/A
Only null left
 
Description

Description

Molybdenite, naturally occurring mineral, grains, is used for grain mounts, XRD and microprobe standards, as well as identification of unknown minerals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1309-56-4
160.07
CWQXQMHSOZUFJS-UHFFFAOYSA-N
14823
bis(sulfanylidene)molybdenum
MoS2
MFCD00003470
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
CHEBI:30704
S=[Mo]=S

Specifications

1309-56-4
MoS2
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
CWQXQMHSOZUFJS-UHFFFAOYSA-N
bis(sulfanylidene)molybdenum
14823
160.07
Molybdenite
1 g
MFCD00003470
Soluble in sulfuric and nitric acid
S=[Mo]=S
160.07
CHEBI:30704
Glass bottle
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-172-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only