missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads

CAS: 63231-69-6

$63.90 - $497.82

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0608530
View Documents
Thermo Scientific Chemicals
L0608530
250 g
Each for $63.90
Only null left
 
AAL060850B
View Documents
Thermo Scientific Chemicals
L060850B
1000 g
Each for $168.75
Only null left
 
AAL060850I
View Documents
Thermo Scientific Chemicals
L060850I
5000 g
Each for $497.82
Only null left
 
Description

Description

Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63231-69-6
MFCD00131613
Molecular sieves 13X

Specifications

Hygroscopic
63231-69-6
Insoluble in water.
Molecular sieves, 13X, 3 to 5mm (0.12 to 0.20 in) beads
250 g
MFCD00131613
Molecular sieves 13X
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only