missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads

CAS: 63231-69-6

Supplier:  Thermo Scientific Chemicals L060850I

 View more versions of this product

Catalog No. AAL060850I


Only null left
Explore available promotions

Description

Description

Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hygroscopic
Molecular sieves, 13X
63231-69-6
Insoluble in water.
5000 g
3 to 5mm (0.12 to 0.20 in) beads
MFCD00131613
Molecular sieves 13X

Chemical Identifiers

63231-69-6
MFCD00131613
Molecular sieves 13X
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only