Learn More
Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads
CAS: 63231-69-6
Supplier: Thermo Scientific Chemicals L060850B
Description
Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hygroscopic | |
| Molecular sieves, 13X | |
| 63231-69-6 | |
| Insoluble in water. |
| 1000 g | |
| 3 to 5mm (0.12 to 0.20 in) beads | |
| MFCD00131613 | |
| Molecular sieves 13X |
Chemical Identifiers
| 63231-69-6 | |
| MFCD00131613 | |
| Molecular sieves 13X |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only