missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-(and-6)-Carboxy SNARF™-1

Catalog No. C1270
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1270 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1270 Supplier Invitrogen™ Supplier No. C1270

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The cell-impermeant pH indicator has pKa of approximately 7.5, thus is useful for measuring pH changes between pH 7 and pH 8

  • Exhibits significant pH-dependent emission shift from yellow-orange to deep red fluorescence under acidic and basic conditions, respectively
  • This pH dependence allows ratio of fluorescence intensities from dye at two emission wavelengths, typically 580nm and 640nm, to be used for quantitative determinations of pH

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, pH Detection

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Microplate Reader
Product Type pH Indicator
Dye Type pH Indicator
Product Line SNARF
Quantity 1 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.