missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Paclitaxel, Oregon Green™ 488 Conjugate (Oregon Green™ 488 Taxol, Flutax-2)

Catalog No. P22310
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22310 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P22310 Supplier Invitrogen™ Supplier No. P22310

Probe for labeling tubulin filaments in live cells

The fluorescent label on this probe is attached to the 7-carbon of the paclitaxel, a strategy that permits selective binding of the probe to microtubules. Researchers have used similar fluorescent paclitaxel derivatives in a variety of systems.

Cell Analysis, Cell Structure, Cytoskeleton

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Dye Type Oregon Green™ 488
Quantity 100 μg
Product Line Oregon Green
Shipping Condition Room Temperature
Label Type Classic Dyes
Product Type Paclitaxel
Sub Cellular Localization Cytoskeleton, Tubulin

For Research Use Only. Not for use in diagnostic procedures.