missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nile Red
Description
Nile red is used to localize and quantitate lipids, particularly neutral lipid droplets within cells. Nile red is almost nonfluorescent in water and other polar solvents but undergoes fluorescence enhancement and large absorption and emission blue shifts in nonpolar environments (Excitation/emission maxima ∼552/636 nm in methanol).
We recommend dissolving in high-quality anhydrous DMSO or DMF to attain a stock concentration up to 1 mM.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 25 mg |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | Fluorescence Microscope |
| Sub Cellular Localization | Cell Membranes & Lipids, Lysosomes |
| Detection Method | Fluorescence |
| Molecular Formula | C20H18N2O2 |
| Physical Form | Solid |
| Color | Red |
| Product Type | Nile Red Stain |
| Excitation/Emission | 552/636 nm |
For Research Use Only. Not for use in diagnostic procedures.