missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Measure-IT™ Thiol Assay Kit DFS Item

Provides easy and accurate quantitation of thiol

Supplier:  Invitrogen™ M30550

Catalog No. M30550


Only null left
Explore available promotions

Description

Description

  • Linear range of 0.05-5μM thiol, making it up to 400 times more sensitive than colorimetric methods based on Ellman's reagent
  • Assay is performed at room temperature; maximum fluorescence signal is attained in 5 minutes and is stable for at least 1 hour

Biochemical Toxicology Assays, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Drug Discovery and Development, Target-Based ADME/Tox Assays

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Thiol Assay Kit
1 Kit
Measure-iT
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.