missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Guanosine 5'-O-(3-Thiotriphosphate), BODIPY™ FL Thioester, Sodium Salt (BODIPY™ FL GTP-γ-S, Thioester)

Catalog No. G22183
Encompass
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G22183 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G22183 Supplier Invitrogen™ Supplier No. G22183
  • ONLINE EXCLUSIVE PRICE
    $992.00
    $813.44 / Each

    Save $178.56

    18.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorescence may be recovered on binding to G-proteins

BODIPY FL GTP-γ-S binds to GTP-binding proteins. To create this fluorescent nucleotide, the BODIPY FL fluorophore was linked through the γ-thiol of GTP-γ-S. The fluorescence of BODIPY FL GTP-γ-S is quenched approx. 90% relative to that of the free dye, but may be recovered on binding to G-proteins. In addition to its potential use for binding studies, the BODIPY FL GTP-γ-S thioether is an important new substrate for Fhit, a member of the histidine triad superfamily of nucleotide-binding proteins.

Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation

Order Info

Shipping Condition: Wet ice

Specifications

Product Type Guanosine 5'-O
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Labeling Method Chemical Labeling, Direct Labeling
Shipping Condition Wet Ice
Concentration 5 mM
Label or Dye BODIPY™ FL
Quantity 50 μL
Product Line BODIPY

For Research Use Only. Not for use in diagnostic procedures.