Learn More
Invitrogen™ EnzChek™ Cellulase Substrate, blue fluorescent, 339/452
Description
The EnzChek™ cellulase substrate was developed for simple and rapid quantitation of cellulase. The assay can be completed in 30 minutes or less. Simply mix the enzyme with the EnxChek™ cellulase substrate working solution, incubate, and read the results using excitation/emission maxima ∼339/452 nm. In contrast to other more complex, multistep assays, the EnzChek™ cellulase substrate is perfect for use in a high-throughput environment. This fluorescence substrate is highly sensitive, with a detection limit as low as 40 μU/mL cellulase. It is also possible to use this substrate in a colorimetric assay, albeit with reduced sensitivity.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| Form | Solid |
| For Use With (Application) | Enzyme and Metabolic Activity |
| Product Line | EnzChek |
| Quantity | 1 mg |
| Substrate | Cellulase substrate |
| Substrate Properties | Chemical Substrate |
| For Use With (Equipment) | Fluorometer, Microplate Reader |
| Shipping Condition | Room Temperature |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.