missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNP-BSA (Albumin from Bovine Serum (BSA), 2,4-Dinitrophenylated)
Description
- Some applications of tracer include study of Fc receptor-mediated immune function and IgE- and IgG-mediated responses to crosslinking of DNP-specific antibodies
- Because DNP-BSA has more defined molecular weight than polydisperse dextrans, it may be better suited for some applications where size of tracer protein must be known with accuracy
- Has approximately 25 dinitrophenyl substituents per molecule of BSA
- DNP-BSA can be detected with unlabeled or fluorescently labeled anti-DNP antibody
Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature
Spécifications
Spécifications
| Quantity | 5 mg |
| Conjugate | DNP (Dinitrophenyl) |
| Species | Bovine |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Other Label(s) or Dye(s) |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Albumin Proteins |
For Research Use Only. Not for use in diagnostic procedures.